Learn More
HSD11B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10928-1-AP-150UL
 View more versions of this product
Product Code. 16837643
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
HSD11B1 is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, HSD11B1 can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children.The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
HSD11B1
Polyclonal
Unconjugated
HSD11B1
11 beta HSD1, 11 DH, HDL, HSD11, HSD11B, HSD11B1, HSD11L, SDR26C1
Rabbit
Antigen Affinity Chromatography
RUO
15483, 25116, 3290
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16232, P28845, P50172
HSD11B1
HSD11B1 Fusion Protein Ag1361
150 μL
Primary
Human, Mouse, Rat