missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HSBP1 (Human) Recombinant Protein
Human HSBP1 full-length ORF ( AAH07515, 1 a.a. - 76 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH07515 |
---|---|
Gene ID (Entrez) | 3281 |
Name | heat shock factor binding protein 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126401
|
Abnova™
H00003281-P01.10ug |
10 μg |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16136401
|
Abnova™
H00003281-P01.25ug |
25 μg |
6245.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sequence: MAETDPKTVQDLTSVVQTLLQQMQDKFQTMSDQIIGRIDDMSSRIDDLEKNIADLMTQAGVEELESENKIPATQKS
Specifications
AAH07515 | |
heat shock factor binding protein 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
DKFZp686D1664, DKFZp686O24200, NPC-A-13 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3281 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSBP1 | |
GST |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein