missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRSP12 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12930-1-AP-20UL
This item is not returnable.
View return policy
Description
Product No. 12930-1-AP is a rabbit polyclonal antibody raised from human full-length UK114.
Endoribonuclease responsible for the inhibition of the translation by cleaving mRNA. Inhibits cell-free protein synthesis. Cleaves phosphodiester bonds only in single-stranded RNA.Specifications
HRSP12 | |
Polyclonal | |
Unconjugated | |
RIDA | |
heat responsive protein 12, HRSP12, P14.5, PSP, Ribonuclease UK114, UK114, UK114 antigen homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10247, 65151 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52758, P52759 | |
Rida | |
HRSP12 Fusion Protein Ag3628 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction