Learn More
Grace Davison™ HPLC Column Visionht™
Brand: Grace Davison™ 5141909
This item is not returnable.
View return policy
Description
- Maximise speed and resolution
- High throughput technologies for all systems
- Simplify separation strategies
- Complementary phase chemistries
- Improve laboratory efficiency
- Seamless method transfer between UHPLC, HPLC and Prep
- UHPLC columns rated to 16,000psi for compatibility on all systems
- Low dead volume column hardware used for all UHPLC columns
- UltraMD UHPLC method development kits available containing C18 classic, C18 polar, C18 high load and C18 basic columns in selected dimension
- Other sizes available
Product Content Correction
Product Title
Grace Davison™ HPLC Column Visionht™ > UHPLC C18 Basic 120A small molecule 100mm x 2mm particle size 1.5μm Grace Discovery Sciences