Learn More
Horst™ Hotbands
Brand:  Horst™ SQ00006
 View more versions of this product
Product Code. 11912033
  • 3470.00 NOK / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Specifications
1.5m
230V
66w
1.5m
230V Voltage
Hotband
Product Title
Horst™ Hotbands > Voltage: 230V; Watts: 66w