Learn More
Honeywell™ Protégé™ Safety Glasses
Brand: Sperian Protection 1015364
This item is not returnable.
View return policy
Description
Specifications
EN 166 and EN 170 1F 3-1.2 | |
Clear | |
General use and protection against impact (45m/s), Well suited for applications such as utilities, light & heavy manufacturing, bio tech, ship, building, construction and automotive manufacturing. | |
1 Ea. | |
Plastic |
Scratch-resistant | |
Eye and Face Protection Spectacles | |
Metallic Black | |
Polycarbonate |