Learn More
Sperian™ Honeywell™ POLYSAFE™ Clear Uncoated Safety Glasses
Click to view available options
Frame Color:
Clear
Description
- Good for mechanical work involving grinding, turning and milling
- Wraparound spectacles for clear and distortion-free vision
- Indirect vents help prevent fogging
- Perforated temples to allow the addition of a safety cord
- Provides 99.9% UV protection
Specifications
Specifications
Certifications/Compliance | EN 166 1F, EN 170 and 2-1.2 |
Lens Tint | Clear |
Product Type | Eye and Face Protection Spectacles |
For Use With (Application) | Fits over prescription Glasses, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling |
Frame Color | Clear |
No. per Pack | 10/Pk. |
Lens Material | Polycarbonate |
Product Content Correction
Product Title
Sperian™ Honeywell™ POLYSAFE™ Clear Uncoated Safety Glasses >