Learn More
Sperian™ Honeywell™ Premium Disposable Respirators
Highly comfortable and ergonomically designed single-use respirators with no restriction with regard to use. Sperian™ Honeywell™ Premium Disposable Respirators leverage the Willtech™ soft seal technology for an improved fit and exceptional comfort.
447.00 NOK - 781.00 NOK
Specifications
Certifications/Compliance | EN 149:2001A1:2009, ISO 9001 / 2000, EC |
---|---|
Disposable | Single-use |
Product Type | Respiratory Protection |
Size | Medium |
Latex Free | Yes |
Product Code | Brand | Filter Type | With Exhalation Valve | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | With Exhalation Valve | Price | Quantity & Availability | ||||
11816960
|
Sperian™
1005580 |
FFP1 | No |
447.00 NOK
Pack of 20 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||||
11806970
|
Sperian™
1005586 |
FFP2 | Yes | N/A | |||||
Description
- Rigid shell so shape is maintained even in moist and heavy loading environments
- Hyopallergenic flange seal with absorption properties
- Preformed and colored nose bridge to save time
- Large internal volume to easily communicate and breathe
- Twin elastic straps, ultra flexible and resistant, stapled outside of the filtering area
- No latex, no silicone, no PVC
- Protection and wearer acceptance guaranteed during periods of extended use
Specifications
EN 149:2001A1:2009, ISO 9001 / 2000, EC | |
Respiratory Protection | |
Yes | |
Dusts, Mists, Fumes |
Single-use | |
Medium | |
20/Pk. | |
Molded Cup |