Learn More
Sperian™ Honeywell™ 4000 Premium Series Respirators
Reduces the risk of contamination and leakage, prevents irritation and allergies and increases safety. Sperian™ Honeywell™ 4000 Premium Series Respirators are easy to store and transport.
Brand: Sperian™ 1005630
This item is not returnable.
View return policy
Description
- Individually packaged, single use
- With ultra-soft hypoallergenic Willtech™ flange seal
- With absorption properties and a tight sealing film for exceptional comfort and fit for long periods
- Preformed and colored adjustable nose bridge
- Vast interior with a built-in chin piece
- Ultra-lightweight with a fluid resistant outer shell
- Sliding straps, highly flexible
Specifications
EN 149:2001A1:2009, ISO 9001 / 2000, EC | |
FFP3 | |
Medium | |
Textile, mining, construction, iron and steel industries, car body preparation industry, woodworking, agricultural and horticultural activities, food industry, cutting and molding of Metals, pharmaceutical manufacturing, nuclear industry, toxic waste treatment, battery, biological risks, etc | |
4311 ML Respirator | |
Dusts, Mists, Fumes |
Single-use | |
Respiratory Protection | |
Yes | |
Yes | |
10/Pk. | |
Flat Fold |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
Sperian™ Honeywell™ 4000 Premium Series Respirators > Single elastic head strap