Learn More
Bochem™ HOFFMANN™ Pinch Cock Tube Clamps
Pinch cock tube clamps
Brand:  Bochem™ 12531
 View more versions of this product
Product Code. 11885091
  • 615.00 NOK / Pack of 10
Only null left
Estimated Shipment: 22-07-2025
Description
  • Pinch cock type
  • Includes flap
Specifications
Specifications
10/Pk.
Ms-Ni
17 mm
Pinchcock Tube Clamp
For pipes up to O.D. 40 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ HOFFMANN™ Pinch Cock Tube Clamps > Width: 17mm