Learn More
Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
Brand: Hoefer™ SUB15-C1-20
This item is not returnable.
View return policy
Description
- Ranges from 10, 16, 20, 25 and 30 Wells
- 1, 1.5 and 2 mm Thickness
- 2.5 to 12.8 mm Well Width
- Min. 12, max. 128 μL Sample Volume
Specifications
SUB15 Standard Submarine | |
20 | |
0.1 cm | |
0.23 in. | |
0.19 in. |
Comb | |
0.03 in. | |
29 μL | |
0.59 cm | |
0.5 cm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
Hoefer™ SUB15 Standard Submarine Combs > Comb: 1mm x 20 Wells; 29μL