Learn More
Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
Brand:  Hoefer™ SUB15-C1.5-25
 View more versions of this product
Product Code. 12635756
  • 905.00 NOK / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Description
  • Ranges from 10, 16, 20, 25 and 30 Wells
  • 1, 1.5 and 2 mm Thickness
  • 2.5 to 12.8 mm Well Width
  • Min. 12, max. 128 μL Sample Volume
Specifications
Specifications
SUB15 Standard Submarine
25
0.15 cm
0.13 in.
0.19 in.
Comb
0.05 in.
25 μL
0.35 cm
0.5 cm
Documents
One moment while we fetch your results.
Product Title
Hoefer™ SUB15 Standard Submarine Combs > Comb: 1.5mm x 25 Wells; 25μL