Learn More
Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
Brand:  Hoefer™ SUB15-C1.5-16MC
 View more versions of this product
Product Code. 12605770
  • 905.00 NOK / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Description
  • Ranges from 10, 16, 20, 25 and 30 Wells
  • 1, 1.5 and 2 mm Thickness
  • 2.5 to 12.8 mm Well Width
  • Min. 12, max. 128 μL Sample Volume
Specifications
Specifications
SUB15 Standard Submarine
16
0.15 cm
0.25 in.
0.19 in.
Comb
0.05 in.
48 μL
0.64 cm
0.5 cm
Product Title
Hoefer™ SUB15 Standard Submarine Combs > Comb: 1.5mm x 16 Wells; 48μL