Learn More
Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates
Use with the SE 250 vertical electrophoresis unit. Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
Brand: Hoefer™ SE202N
This item is not returnable.
View return policy
Description
- 10 x 8 cm
Specifications
SE 250 Vertical Electrophoresis Unit | |
Notched Alumina Plate | |
Alumina | |
1/Pk. |
Product Content Correction
Product Title
Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates > Alumina plate; Notched; 10 x 8cm