missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA ABC Antibody (MHC-I/8147R), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20852AF647
This item is not returnable.
View return policy
Description
HLA ABC Monoclonal antibody specifically detects HLA ABC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
HLA ABC | |
Monoclonal | |
Alexa Fluor 647 | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Recombinant fragment (around aa100-300) of human HLA-I protein (exact sequence is proprietary) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology | |
3105 | |
Store at 4°C in the dark. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
MHC-I/8147R | |
50 mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HLA ABC Antibody (MHC-I/8147R), Alexa Fluor™ 647, Novus Biologicals™ > 0.1 mL, Alexa Fluor 647
Spot an opportunity for improvement?Share a Content Correction