Learn More
Simport™ Scientific Histosette™ II Biopsy Processing/Embedding Cassette in Quickload™ Stacks
Secure specimens during processing with Simport Scientific™ Histosette™ II Biopsy Processing/Embedding Cassette. Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Description
In Quickload™ sleeve
- 45° anterior writing area
- Open and close as often as necessary — cassettes will always relock securely
- Back mounted tab allowing an unobstructed anterior marking area
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
| Acetal | |
| 2000 |