missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1860.00 NOK - 4380.00 NOK
Specifications
Antigen | Histone H2AX |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329792
|
Bio-Techne
NBP3-05219-20UL |
20 μL |
1860.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18394152
|
Bio-Techne
NBP3-05219-100UL |
100 μL |
4380.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Histone H2AX | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
3014 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific | |
PBS with 50% glycerol, pH7.3. | |
Recombinant protein of human Histone H2AX. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title