missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen Capto Butyl ImpRes

HiScreen Capto Butyl ImpRes is a ready-to-use column prepacked with Capto Butyl ImpRes hydrophobic interaction chromatography resin and is part of Cytiva’s process development platform.

Brand:  Cytiva 17371910


Product Code. 15599261

  • 2190.00 NOK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Ready-to-use column prepacked with Capto Butyl ImpRes hydrophobic interaction chromatography resin
  • Part of Cytiva’s process development platform
TRUSTED_SUSTAINABILITY
Specifications

Specifications

20% ethanol at 4 to 30 °C
100 mm
4.7 mL
7.7 mm
3 bar/0.3 MPa/44 psi
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HiScreen Capto Butyl ImpRes > 4.7 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.