Learn More
Nickel Electro™ Hinged Gable Lid
View samples better with a semi-transparent lid. Nickel Electro™ Hinged Gable Lid maximizes bath area while using its shape to drip condensation back into the bath.
Brand:  Nickel Electro™ GL1-4
 View more versions of this product
Product Code. 11419519
  • 1175.00 NOK / Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
Includes: dimForUseWithEquipment
Requires : ForUseWithApplication
Description
Description
  • Lightweight
  • Semi-transparent
  • Drips condensation back into bath
Transparent lid, hinged, high profile, handle
Specifications
Specifications
2 L and 4 L Unstirred Baths
Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Nickel Electro™ Hinged Gable Lid > For Use With: 2 and 4L Unstirred Baths