Learn More
Julabo™ HighTech Heating Circulators
Julabo™ HighTech Heating Circulators features stainless steel bath tanks that are suitable for both internal and external temperature control.
Brand:  Julabo™ 9252526
 View more versions of this product
Product Code. 11974808
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • External temperature control in closed and open systems
  • ICC temperature control for high precision
  • VFD Comfort Display with simultaneous display of 3 temperature values
  • Integrated programmer with real time clock
  • Powerful pressure and suction pump, electronically adjustable
  • Automatic adjustment of pump capacity to fluid viscosity
  • RS232 interface for online communication
  • Integrated cooling coil
Specifications
Specifications
26 L
22 to 26 L/min.
Heating Circulator
2 Barbed fittings for tubing 8 and 12mm I.D. (pump connections M16 x 1 female)
SE-26
61 x 36 x 47 cm
20 cm
230 V 50/60 Hz 12/13 A
Classification III (FL) according to DIN 12876-1
20°C to 300°C
3 kW
0.4 to 0.7 bar
±0.01°C
22 cm
27 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Julabo™ HighTech Heating Circulators > VFD display; Capacity: 26L