Learn More
Fisherbrand™ High Speed Module
High speed module consist of 2 tanks on top of the machine.
Brand:  Fisherbrand™ 2918421
 View more versions of this product
Product Code. 17147844
  • 86110.00 NOK / Each
Estimated Shipment: 07-08-2025
Description
  • Tanks electrically heated
  • Water is preheated before going into the washing chamber, allowing for shorter cycle times
Specifications
Specifications
High Speed Module
Blue Line 6 / IQ6
2 Tanks
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ High Speed Module > Blue Line 6 / IQ6