Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Floor Stands
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Heraguard™ ECO clean bench.
Brand:  Thermo Scientific™ 50109313
 View more versions of this product
Product Code. 10690394
  • 16460.18 NOK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Description
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Heraguard ECO 1.8, 1.8/95
Provides variable ECO work height of 760 mm - 960 mm in 50 mm adjustable increments.
Adjustable Floor Stand
680 to 880 mm
SDS
Documents Certificates
View accessory products
Promotions
Promotions
  • Multiple Manufacturers
    Save big on essential products for cancer research
    Promo Code24147
    Expires2025-07-31
    New Offer
    Save Now
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Heraguard™ ECO Clean Bench Floor Stands > Adjustable floor stand 1.8m, provides variable ECO work height of 760mm - 960mm in 50mm adjustable increments.