Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Floor Stands
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Heraguard™ ECO clean bench.
Brand: Thermo Scientific™ 50109311
Description
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
†Due to DC motors, HERAguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
Heraguard ECO 1.8, 1.8/95 | |
Provides ECO Work Height of 760 mm | |
Floor Stand | |
680 mm |
Promotions
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%