Learn More
BRAND™ Hellendahl Staining Trough
Soda-lime glass construction, for 16 slides
Brand:  BRAND™ 472700
3263.96 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15603977
  • 4140.00 NOK / Pack of 10
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 07-07-2025
Description
  • For 16 slides, size 76 x 26 mm
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    16
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    BRAND™ Hellendahl Staining Trough > Dimensions (L x W): 60 x 55 mm