Learn More
Heidolph™ Hei-VAP Expert Control ML G6B
Adds numerous smart features to the Hei-VAP Expert, as well as an overview of all parameters and the current process.
Brand:  Heidolph™ 574-02610-00
76371.84 NOK valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 26.00000kg
Product Code. 16400044
  • 85970.00 NOK / Each
Estimated Shipment: 06-08-2024
Description
  • 5 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Motor lift model with transparent plastic coated G6 vertical glassware for reflux distillation
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • With USB and Micro SD interface for easy installation of software updates and transfer of individual favorites from one device to another
Specifications
Specifications
EU Plug
Motor Lift Rotary Evaporator