Learn More
Heidolph™ Hei-VAP Expert Control HL G3
Adds numerous smart features to the Hei-VAP Expert, as well as an overview of all parameters and the current process.
Brand: Heidolph™ 573-02300-00
53088.01 NOK valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- 5 in. touch display for controlling the entire system including the vacuum and the cooling temperature
- Hand lift model with G3 vertical glassware
- For precise monitoring, the pictogram on the display also symbolizes active functions
- Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
- Setting of values either via the touch display or the separate quick access knobs
- Direct access to two favorites on the start screen
- Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
- Define and save own evaporation processes (ramps)
- Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
- With USB and Micro SD interface for easy installation of software updates and transfer of individual favorites from one device to another
Specifications
EU Plug | |
Hand Lift Rotary Evaporator |