Learn More
3M™ Heavy Duty QRS Breathing Tube Product Code.: 15719767
Only undefined left
Add to basket
3M™ Heavy Duty QRS Breathing Tube
Product Code. 15719767
Color: Black
Click to view available options
:
Color: Black
Product Code. 15719767
Brand: 3M™ 7000044527
Only undefined left
Add to basket
Only undefined left
Add to basket
3M™ Heavy Duty QRS Breathing Tube for 3M™ Adflo™ Powered Air Respirator and 3M™ Versaflo™ Supplied Air Regulator V-500E.
  • The new welding helmets and faceshields from 3M feature the QRS type of breathing tube (Quick Release Swivel), which enables one-handed connection and release
  • The protection class of Speedglas welding helmet 9100FX Air is reduced to TH2 when the adapter is used
  • You can extend the service life of the breathing tube by using a tube cover
TRUSTED_SUSTAINABILITY
Specifications
Material Heavy Duty Rubber
Product Content Correction
Product Title
3M™ Heavy Duty QRS Breathing Tube >