Learn More
Medline Scientific™ Heating Mantles
Brand:  Medline Scientific™ ES303
 View more versions of this product
Product Code. 10484622
  • 13575.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 29-08-2025
Specifications
500 mL
Heating Mantle
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Medline Scientific™ Heating Mantles > Capacity: 500mL