Learn More
Eppendorf™ Heat Sealer S100
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
37956.22 NOK
Specifications
Hertz | 50/60 Hz |
---|---|
Voltage | 100/230 V |
Dimensions (L x W x H) | 220 × 250 × 230 mm |
Electrical Requirements | 100/230 V, 50/60 Hz |
For Use With (Equipment) | 96- and 384-well plates |
Description
- Hermetic sealing of multiwell plates
- Fixed sealing temperature for reproducible results
- Eliminates evaporation in PCR, reducing cross contamination
- Seals 96- and 384-well plates
- Compact design, portable device
- Integrated thermostats prevents overheating
- Improved mechanics for even easier sealing
- PCR plate adapter included
Specifications
50/60 Hz | |
220 × 250 × 230 mm | |
96- and 384-well plates |
100/230 V | |
100/230 V, 50/60 Hz |
Product Title
Eppendorf™ Heat Sealer S100