Learn More
Kartell™ HDPE Stackable Tank
Brand:  Kartell™ 0060200
Product Code. 15676395
  • 424.00 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Specifications
HDPE
20 L
200 mm
1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ HDPE Stackable Tank > Capacity: 20L