Learn More
Horst™ HBST Model Heating Tape
Brand:  Horst™ 20704
 View more versions of this product
Product Code. 10077081
  • 2440.00 NOK / Each
Only null left
Estimated Shipment: 18-06-2025
Specifications
4 Lm
4 m
HBST
4 Lm
∼230V Voltage
Heating Tape
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Horst™ HBST Model Heating Tape > Length: 4m