missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Woodsafe Hazardous Waste Bin - Sharpsbin
A cost-effective solution for lowering the CO2 emissions. Eco-friendly sharps and hazardous waste containers, produced from renewable resources using green energy. Reducing CO2 emissions by up to 66%, documented in a full LCA. Available in sizes from 0.5 L to 50 L.
27652.27 NOK - 31172.71 NOK
Specifications
Certifications/Compliance | EN ISO 23907-1:2019. UN3291, ADR Approved |
---|---|
Closure Type | Temporary and permanent closure |
Closure Color | Brown |
Description | Environmental friendly Sharpsbin |
Color | Brown, Yellow |
Alert:
Follow instructions for safe and correct use
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
18792815
|
Woodsafe
1250120 |
25 L |
27652.27 NOK
Pack of 132 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772825
|
Woodsafe
1500120 |
50 L |
31172.71 NOK
Pack of 96 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry.
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Made from 100% Biocomposite
- The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal.
- Natural colour and smell from wood - No migration of substances - EN - tested for toys and food
- Compliant with UN3291
- Tested according to ISO 23907-1:2019
- UN ADR Approved
- Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
- NOTE: String between lids is for delivery purpose, remove/break before use!
- Stop burning plastic
Specifications
EN ISO 23907-1:2019. UN3291, ADR Approved | |
Brown | |
Brown, Yellow | |
Yes | |
Autoclavable | |
Safe Disposal of sharps and potentially Infectious Waste or medical waste | |
Biocomposite |
Temporary and permanent closure | |
Environmental friendly Sharpsbin | |
Single-use | |
Non-sterile | |
Bin + Lid + Label | |
Pallet | |
Biohazard Waste Container (Benchtop) |
Lower climate impact
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title