Learn More
Sartorius™ Handle
Brand:  Sartorius™ YAW52
4632.72 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10663922
  • 4755.00 NOK / Each
Specifications
Handle
20kg Weights
1 Each
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Handle > For Use with 20kg Weights