Learn More
Sartorius™ Handle
Brand:  Sartorius™ YAW51
3650.02 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10367251
  • 3750.00 NOK / Each
Only null left
Estimated Shipment: 22-07-2025
Specifications
Handle
10kg Weights
1 Each
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Handle > For Use with 10kg Weights