Learn More
Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System
A complete set of tools for aspiration—rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes with the Argos Technologies HandE-Vac Handheld Aspirator.
3290.00 NOK - 7075.00 NOK
Specifications
Compatible Tips | E-Vac Only |
---|---|
Adjustable Tip Spacing | No |
Autoclavable | Autoclavable |
Color | Black |
Disposable | Reusable |
Product Code | Brand | Number of Channels | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Number of Channels | Price | Quantity & Availability | ||||||
15944991
|
Cole-Parmer™ Essentials
EV514 |
1 |
3290.00 NOK
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
15934991
|
Cole-Parmer™ Essentials
EV500 |
1 or 8 | N/A | |||||||
Description
- Use with Pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications
- Pressure-sensitive button on the top of the unit controls the specific amount of vacuum appliedOperates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems
- Autoclavable
Specifications
E-Vac Only | |
Autoclavable | |
Reusable | |
Plastic | |
Handheld Aspirator |
No | |
Black | |
Ergonomic | |
HandE-Vac | |
4 L |
Product Title
Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System