Learn More
JG Finneran Associates™ Hand Operated Vial Decapper
3180.00 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
12493030
|
JG Finneran Associates™
9320-11 |
11 mm Crimp Vials |
3180.00 NOK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
12483030
|
JG Finneran Associates™
9320-08 |
8 mm Crimp Vials |
3180.00 NOK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
Specifications
Vial Decapper | |
1 Ea. |
Product Title
JG Finneran Associates™ Hand Operated Vial Decapper