Learn More
Hanna Instruments™ Hand Conductivity Meter
Brand: Hanna Instruments™ HI8733
7177.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Manually via trimmer. | |
Hand Conductivity Meter | |
Approximately 100 hr. of continuous use | |
9 V |
Product Content Correction
Product Title
Hanna Instruments™ Hand Conductivity Meter > Voltage: 9V