Learn More
Hamilton™ 800 Series Microliter™ Replacement Syringe Barrels
Barrels available with or without needle. Choose removable needle (RN) or cemented needle (N) termination. (See exact specifications below.)
Brand:  Hamilton™ 32151
 View more versions of this product
Product Code. 10007561
  • 1105.00 NOK / Each
Explore more special offers
Specifications
Liquids
Barrels/Plunger Assembly with Needle
801 N Syringe
With needle (26s/51/2)
10 μL
Product Title
Hamilton™ 800 Series Microliter™ Replacement Syringe Barrels > 801 N