missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamartin/TSC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | Hamartin/TSC1 |
---|---|
Concentration | 600 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17629706
|
Proteintech
29906-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17619706
|
Proteintech
29906-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hamartin/TSC1 Polyclonal antibody apecifically detetects Hamartin/TSC1 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
Hamartin/TSC1 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
7248 | |
IgG | |
Antigen affinity purification |
600 μg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse | |
NM_000368 | |
TSC1 | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title