Learn More
Gilac™ Half Size Dough Container
Click to view available options
Volume (Metric):
9 L
Unit Size:
Each
Description
- 400 x 300 dough storage container.
- Stacking with and without lid, can be stacked to a great height
- Easy cleaning flat bottom.
Specifications
Specifications
| Product Type | Dough Container |
| Material | Polyethylene |
| Color | White |
| Dimensions (L x W x H) | 400 x 300 x 100 mm |
| Volume (English) | 2.4 gal. |
| Volume (Metric) | 9 L |
Product Content Correction
Product Title
Gilac™ Half Size Dough Container >