Learn More
CleanSpace™ Half Mask Adaptor Product Code.: 15950052
Only undefined left
Add to basket
CleanSpace™ Half Mask Adaptor
Product Code. 15950052 Shop All CleanSpace Technology Products
Product Type: Half Mask Adaptor
Click to view available options
:
Product Type: Half Mask Adaptor
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15950052
Brand: CleanSpace™ PAF0025
Only undefined left
Add to basket
Only undefined left
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Half mask adaptor is equipped for the front of the mask to enable quantitative fit testing.
  • Used with the revolutionary CleanSpace™ Respirators: light weight, no hoses or belts
  • Used with the CleanSpace™ Half Masks only
  • Quick and easy to install
  • Fits with the TSI PortaCount equipment
  • Easy to wash and quick drying
Specifications
Product Type Half Mask Adapter
For Use With (Application) Quantitative Fit Testing
For Use With (Equipment) PAF-1010, PAF-0027, PAF-0033
Model PAF-0025
Product Content Correction
Product Title
CleanSpace™ Half Mask Adaptor >