Learn More
Eppendorf™ Half Height Freezer Rack for Chest Freezers
Store your freezer storage boxes within organized freezer racks. These metal racks optimize your freezer space and make it easy to find the items you desire.
1636.52 NOK - 1942.74 NOK
Specifications
Product Type | Half Height Freezer Rack |
---|---|
For Use With (Equipment) | Chest Freezer |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
13468676
|
Eppendorf™
K0641-3007 |
pk |
1636.52 NOK
Each |
In Stock
Log in to see stock. |
|||||
13478676
|
Eppendorf™
K0641-306A |
2 in. Boxes (7) |
1942.74 NOK
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
13488676
|
Eppendorf™
K0641-3008 |
4 in. Boxes (3) |
1636.52 NOK
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
Description
- Half Height Freezer Rack for Chest Freezers
- Available in a variety of sizes
Specifications
Half Height Freezer Rack | |
Chest Freezer |