Learn More
GW3965 hydrochloride, MedChemExpress Product Code.: 16406787
GW3965 hydrochloride, MedChemExpress
Product Code. 16406787
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16406787
Brand: Medchem Express HY10627A
MedChemExpress GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
Specifications
Chemical Name or Material GW3965 hydrochloride
CAS 405911-17-3
Color White
Purity Grade Notes Research
Molecular Formula C33H32Cl2F3NO3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (161.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES ClC1=C(C(F)(F)F)C=CC=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.[H]Cl
Molecular Weight (g/mol) 618.51
Formula Weight 618.51
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
GW3965 hydrochloride, MedChemExpress