Learn More
Saint-Gobain GSR Rubber Tubing
Use this tubing for a variety of industrial, construction and chemical applications. Saint Gobain Performance Plastics™ GSR Rubber Tubing provides an ideal combination of chemical resistance and pump life.
Brand:  Saint-Gobain 71011510
 View more versions of this product
Product Code. 11836331
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Very good elasticity and flexibility
  • Good resistance to abrasion
  • 45 shore A natural rubber construction
  • Light, supple and wear-resistant with good impermeability
  • Provides good chemical resistance to acids and bases
  • Meets FDA criteria
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDA compliant, food WCR
Natural Rubber
10 m
General laboratory use
Red
70°C
9 mm
13 mm
2 mm
-25°C
Flexible