Learn More
Tocris Bioscience™ GSK LSD 1 Dihydrochloride
Potent and selective LSD1 inhibitor
Brand: Tocris Bioscience™ 5361/10
This item is not returnable.
View return policy
Description
GSK LSD 1 dihydrochloride is a potent and selective lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 16 nM). Exhibits >1000-fold selectivity for LSD1 over closely related enzymes including LSD2, MAO-1 and MAO-B. Inhibits cancer cell line growth in vitro.Specifications
GSK LSD 1 dihydrochloride | |
rel-N-[(1S,2R)-2-Phenylcyclopropyl]-4-piperidinamine dihydrochloride | |
Soluble to 100mM in water and to 100mM in DMSO | |
Bioactive Small Molecule |
2102933-95-7 | |
10 mg | |
Solid |
Research Use Only. Not For Human or Veterinary Use