missing translation for 'onlineSavingsMsg'
Learn More

GSK J2, Tocris Bioscience™

Product Code. 30002064
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inactive isomer of GSK J1 (Cat. No. 4593)

Inactive control of GSK J1 (IC50 > 100 μM for inhibition of JMJD3/UTX).

Cell permeable ester derivative, GSK J5 and Active Analog also available.

Chemical Identifiers

CAS 1394854-52-4
Molecular Formula C22H22N5O2-
Molecular Weight (g/mol) 388.451
InChI Key LJIFOCRGDDQFJF-UHFFFAOYSA-M
PubChem CID 122173063
IUPAC Name 3-[[2-pyridin-3-yl-6-(1,2,4,5-tetrahydro-3-benzazepin-3-yl)pyrimidin-4-yl]amino]propanoate
SMILES C1CN(CCC2=CC=CC=C21)C3=CC(=NC(=N3)C4=CN=CC=C4)NCCC(=O)[O-]

Specifications

Color White
Quantity 50 mg
Formula Weight 389.45
Percent Purity >98%
Physical Form Powder
Chemical Name or Material GSK J2
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'