missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSDMD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20770-1-AP-20UL
This item is not returnable.
View return policy
Description
FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.Specifications
GSDMD | |
Polyclonal | |
Unconjugated | |
GSDMD | |
DF5L, DFNA5L, FKSG10, gasdermin D, GSDMD, GSDMDC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79792 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P57764 | |
GSDMD | |
GSDMD Fusion Protein Ag14757 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction