Learn More
GSDMB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12885-1-AP-20UL
This item is not returnable.
View return policy
Description
The GSDMB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GSDMB. This antibody recognizes human antigen. The GSDMB antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
GSDMB | |
Polyclonal | |
Unconjugated | |
20 μL | |
55876 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |