Learn More
Retsch™ Grinding Jar
Retsch™ Grinding Jar, when used with the CryoMill, is continually cooled with liquid nitrogen from the integrated cooling system before and during the grinding process.
Brand:  Retsch™ 01.462.0332
 View more versions of this product
Product Code. 15393535
  • 15375.00 NOK / Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
Description
Description
  • Screw-top
  • For convenient leak-proof operation
  • Constructed of stainless steel or hardened steel
  • Performs radial oscillations in a horizontal position
Specifications
Specifications
Grinding Jar
50 mL
CryoMill
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Retsch™ Grinding Jar > Capacity: 50 mL