missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Snap Caps
For recapping 13mm VACUETTE tubes
3250.00 NOK
Specifications
Product Type | Snap Cap |
---|---|
Material | PET |
Size | Universal |
For Use With (Equipment) | Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |
Description
Specifications
Snap Cap | |
PET | |
Universal | |
Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |